Would you like to log in again?
Ward's® Live Staphylococcus epidermidis Culture
Supplier: VWR
WARD470176-542 WARD470176-544
470176-542EA 13.25 USD InStock
470176-542 470176-544
Ward's® Live Staphylococcus epidermidis Culture
Specimens Live Specimens Bacteria - Living Material
This organism is normal flora of human skin.
  • Biosafety Level: 1
  • Gram Stain: Positive
  • Morphology: Cocci in grape-like clusters
  • Coagulase: Negative
  • Catalase: Positive
  • Motility: Negative
  • Respiration: Aerobic
All bacterial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.
This organism is offered as a ready-to-use tube culture, as well as in a freeze-dried culture for longer storage.
Freeze-dried organisms come with everything you need to rehydrate and culture the organism when you're ready.
Ordering information: These products contain living or perishable materials and will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Bacterial and fungal cultures are restricted and can only be ordered by educational and research professionals.
Ward's Science is not affiliated with the American Type Culture Collection and does not provide corresponding ATCC numbers for our organism.
Caution: Though this organism is not considered to be pathogenic, Ward's recommends that you utilize proper aseptic technique when handling all microorganisms.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty