Would you like to log in again?
Live Amoeba proteus cultures
Supplier: Avantor
WARD470180-124 WARD470176-572 WARD470189-294 WARD470176-446 WARD470176-570
470180-124EA 29.95 USD InStock
470180-124 470189-294 470176-570 470176-572 470176-446
Live Amoeba proteus cultures
Specimens Live Specimens Protists - Living Material
Demonstrate amoeba structure, locomotion, and feeding behavior.
  • Demonstrate amoeba structure, locomotion, and feeding behavior
  • ReadySlides are prepared for you you and offer a convenient, ready-to-go option
  • Vital Stained cultures allow students to better visualize organism structures
Amoeba proteus is a great sarcodine for demonstrating amoebae structure. They use pseudopods for both locamotion and feeding. Amoeba feed on Chilomonas, or other small protozoa, which they phagocytize with their pseudopodia. Amoeba cultures are supplied in Amoeba Media with a rice grain and food (Chilomonas).
Ordering information: These products contain living or perishable materials. Please specify a delivery date.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty