Would you like to log in again?
Ward's® Live Stentor coeruleus Culture
Supplier: VWR
WARD470180-164 WARD470176-586 WARD470176-584
470176-584EA 9.5 USD InStock
470176-584 470176-586 470180-164
Ward's® Live Stentor coeruleus Culture
Specimens Live Specimens Protists - Living Material
This organism is easily recognized by its trumpet-shape and blue coloration. It is quite large (approx. 2 mm) and is a good option for students that are just starting to look at the microscopic world.
  • Large and easy for inexperienced students to see
  • Reproduces both sexually and asexually
  • Compare to Vorticella for differences in cilia arrangement
  • Vital-Staining is available for this culture
  • Cytoplasmic elements are stained blue on your live culture to allow for better visibility
  • Skip the wet-mount, this specimen is available as a set of 5 Readyslides
Stentor is a heterotrich, with different cilial structures on different areas of their cell. They have cilia on the rim of the trumpet, for feeding, but they also have them along their body, allowing them to move from place to place.
Ordering information: Cultures feature a macronucleus as well as a micronucleus. The Macronucleus (which looks like a string of beads) regulates asexual reproduction mechanisms, and the micronucleus aids with sexual reproduction, or conjugation. They are typically found attached to substrates, so they usually adhere to the walls and floor of the specimen jar. They feed on bacteria or other protozoa. Genus: Stentor. Species: coeruleus.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty