Would you like to log in again?
Ward's® Live Freshwater Rotifer Culture
Supplier: VWR
WARD470176-592 WARD470176-594
470176-592EA 9.5 USD InStock
470176-592 470176-594
Ward's® Live Freshwater Rotifer Culture
Specimens Live Specimens Aquaria - Living Material
Study the Rotifera under your microscope to discover the variety of structures among them. Subculture them in Alfalfa or Hay medium to increase your supply. Species may vary based on availability.
Rotifers have a wide variety of movement types ranging from free moving to sessile. They can be found in any healthy freshwater lakes and ponds.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty