Would you like to log in again?
Ward's® Live Spirogyra Culture
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
WARD470176-596 WARD470194-676
470176-596EA 29.95 USD InStock
470176-596 470194-676
Ward's® Live Spirogyra Culture
Specimens Live Specimens Protists - Living Material
Spirogyra is a genus of filamentous green algae with chloroplasts arranged in a helical (spiral) pattern.
  • Excellent food source for freshwater snails and fish
  • Undergoes both sexual and asexual reproduction
Spirogyra are a genus of Chlorophyte algae that can be found in freshwater lakes and ponds. They are characterized by green plastids and cell walls composed of cellulose.
Ordering information: Genus: Spirogyra. Species: not specified.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty