Would you like to log in again?
Ward's® Live Chlorella pyrenoidosa Culture
Supplier: VWR
WARD470176-674 WARD470176-676
470176-674EA 9.5 USD InStock
470176-674 470176-676
Ward's® Live Chlorella pyrenoidosa Culture
Specimens Live Specimens Protists - Living Material
This species of freshwater algae is commonly used for research on photosynthesis.
  • Excellent for studying photosynthesis
  • Rich in cellulose
It is characterized by green plastids and cell walls composed of cellulose. This species has a pyrenoid, which is the organelle that acts as the center of carbon dioxide fixation.
Genus: Chlorella Species: pyrenoidosa
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty