Would you like to log in again?
Live Paramecium bursaria
Supplier: CONNECTICUT VALLEY BIO SUPPLY
WARD470176-724 WARD470176-722
470176-722EA 9.5 USD InStock
470176-722 470176-724
Live Paramecium bursaria
Specimens Live Specimens Protists - Living Material
Paramecium bursaria is a ciliate that contains mutualistic algae called Zoochlorella. The algae lives in the cytoplasma of the Paramecium and provides it with food. Paramecium bursaria grows in Dilute Alfalfa Medium (Hay Medium) or Soil Water Medium. Requires light for growth.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty