Would you like to log in again?
Ward's® Live Paramecium multimicronucleatum Culture
Supplier: VWR
WARD470199-682 WARD470176-726
470176-726EA 27.95 USD InStock
470176-726 470199-682
Ward's® Live Paramecium multimicronucleatum Culture
Specimens Live Specimens Protists - Living Material
Paramecium multimicronucleatumis one of the largest species of Paramecium. This ciliate contains a single macronucleus and four or more micronuclei. Grows in Dilute Alfalfa medium (Hay Medium).
Ordering information: These products contain living or perishable materials and will ship via overnight or 2nd day delivery.
Delivery information: Please specify a delivery date during checkout.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty