Would you like to log in again?
Planaria Slides
Supplier: Avantor
Avg. Ratings: 5.0 (1 Review)
REVIEWS (1)
Total Ratings: 1
Avg. Ratings: out of 5
Sort all by:
WARD470176-912 WARD470181-550 WARD470177-518 WARD470181-552
470176-912EA 7.5 USD InStock
470176-912 470177-518 470181-552 470181-550
Planaria Slides
Slides Prepared Slides
Investigate triploblastic animals
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
We offer four individual Planaria slides in whole mount and cross section orientation. Subjects are fed various food sources to demonstrate digestive systems. Visit our website for full details.
Ordering information: Planaria (whole mount), hematoxylin; represents the general structures of typical triploblastic animals. Planaria, Carbon-fed (whole mount), Schneider's; digestive system is filled with carbon, making branches and diverticula visible. Planaria, Plain and Carbon-fed (whole mount), Schneider's and hematoxylin; Comparitive slide of both a plain and carbon-fed animal, side by side. Planaria, three regions (cross section), hematoxylin and triosin; Composite, illustrates the triclad gastro vascular cavity of flat worms. Anterior, middle, and posterior regions are represented.
Order Now
ORDER
ACCESSORIES
WRITE A REVIEW
REVIEWS (1)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty