Would you like to log in again?
Insect Wings, Composite Slide
Supplier: Avantor
WARD470176-946
470176-946EA 18.5 USD InStock
470176-946
Insect Wings, Composite Slide
Slides Prepared Slides
Compare and Contrast Wing Types.
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Illustrates four types: membranous (housefly), net-veined (dragonfly), scaled (butterfly), and leathery (grasshopper). Unstained.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty