Would you like to log in again?
Animal Mitosis, Parascaris Uterus Slide
Supplier: Avantor
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
WARD470176-948
470176-948EA 16.5 USD InStock
470176-948
Animal Mitosis, Parascaris Uterus Slide
Slides Prepared Slides
Examine cellular division
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Parascaris uterus slide showing diploid chromosomes, centrosomes and spindles.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty