Would you like to log in again?
Ward's® Peranema
Supplier: VWR
WARD470177-302
470177-302EA 9.5 USD InStock
470177-302
Ward's® Peranema
Specimens Live Specimens Protists - Living Material
Extremely small euglenoid. Usually doesn’t rotate when swimming. Scavenger. Hay medium.
Characterized by flagella; lacks plastids.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty