Would you like to log in again?
Crossing Over in Sordaria Slide
Supplier: Avantor
WARD470177-436
470177-436EA 26.25 USD InStock
470177-436
Crossing Over in Sordaria Slide
Slides Prepared Slides
Whole mount
  • All identifying features are clearly distinguishable
  • Rigorous quality control standards
Shows a wild-type and a mutant cross. The arrangement of ascospores in the asci are clearly displayed and are used to estimate the percentage of crossing over between the centromere and the gene that controls the tan spore color.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty