Would you like to log in again?
Human Blood Slide
Supplier: Avantor
WARD470177-634
470177-634EA 6.95 USD InStock
470177-634
Human Blood Slide
Slides Prepared Slides
Investigate the components within your blood
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Human. Shows the three major functional classes: red cells, white cells and platelets. Human red blood cells have no nuclei.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty