Would you like to log in again?
Introductory Nerve Tissues Slide Set
Supplier: Avantor
WARD470177-708
470177-708KT 99 USD InStock
470177-708
Introductory Nerve Tissues Slide Set
Slides Prepared Slides
Introduction to the Various Types of Excitable Cells
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Introduce your students to the nervous system and the many different components involved within this biological network.
Ordering information: Set includes Motor Neurons: Giant multipolar motor neurons; neuron connecting to muscle fibers (sm); Nissl bodies in neurocytes (sect); Glial Cells: Neuroglia, protoplasmic astrocytes (sect); Medullated Nerves: Medullated nerve fibers, teased peripheral nerves (wm); Medullated nerve (cs/ls); Sensory Neurons: Pacinian corpuscle, pressure (cs); Neuro-epithelium (sect); Spinal Cord; Spinal cord (cs); Spinal ganglion (ls); Sympathetic ganglion (ls).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty