Would you like to log in again?
Bottles, HDPE, Narrow Mouth, with Screw Cap
Supplier: Dynalon
301715-0004 414004-157 414004-159 414004-158 2004-0008 2004-0002
470191-304CS 220.3 USD InStock
470191-304 470191-316 470178-134 470191-318 470211-760 470211-762
Bottles, HDPE, Narrow Mouth, with Screw Cap
Bottles Plastic Bottles
Unbreakable and Leak-proof.
  • Each bottle has screw cap with protective inner lining
  • Use amber bottles for light sensitive liquids
These polyethylene bottles are resistant to most acids, bases, alcohols, aliphatic hydrocarbons, and ketones.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty