Would you like to log in again?
Ward's T4rIIB Phage
Supplier: VWR
470179-034EA 18.95 USD InStock
470179-034
Ward's T4rIIB Phage
Specimens Live Specimens Viruses - Living Material
Point mutation in cistron B; pseudoallele of T4rIIA. Reversion rate to wild type approximately 106.
The liquid coliphage suspension is routinely tested for quality, purity, and titer. It contains DNA or RNA supplied as a filtered tryptic soy broth suspension of relative infective titer (108 plaque-forming units per mL). It comes with culture information. The liquid suspension is in tryptic soy broth. It is supplied in a vial and the proven titer is noted on the label. Quantity: 3 mL. Keep refrigerated.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty