Would you like to log in again?
Ward's® Live Escherichia coli DH5 alpha (DH5α) Culture
Supplier: VWR
WARD470179-046
470179-046EA 13.25 USD InStock
470179-046
Ward's® Live Escherichia coli DH5 alpha (DH5α) Culture
Specimens Live Specimens Bacteria - Living Material
This non-pathogenic strain of E. coli was initially created for cloning use.
  • Biosafety Level: 1
  • Gram Stain: Negative
  • Morphology: Rods
  • Respiration: Aerobic
Ordering information: Microbial and fungal cultures are restricted and can only be ordered by educational professionals.
These products contain living or perishable materials and will ship via overnight or second day delivery. Please specify a delivery date during checkout
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty