Would you like to log in again?
Ward's® Live Escherichia coli JM101 (Host) Culture
Supplier: VWR
WARD470179-048
470179-048EA 12.95 USD InStock
470179-048
Ward's® Live Escherichia coli JM101 (Host) Culture
Specimens Live Specimens Bacteria - Living Material
E. coli with JM101 can be transformed with pUC8 plasmid to become resistant to Ampicillin and to metabolize lactose
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty