Would you like to log in again?
Ward's® Live Escherichia coli (non-muccoid) Culture
Supplier: VWR
WARD470179-066
470179-066EA 13.25 USD InStock
470179-066
Ward's® Live Escherichia coli (non-muccoid) Culture
Specimens Live Specimens Bacteria - Living Material
Used for culturing Dictyostelium discoideum (cellular slime mold). 
  • Biosafety Level: 1
  • Canada: Containment Level 2 - PHAC Licence required
  • Gram Reaction: Negative
  • Motility: Positive
  • Respiration: Aerobic
  • Lactose Fermentation: Positive
Ordering information: These products contain living or perishable materials and will ship via overnight or second day delivery. Please specify a delivery date during checkout
Bacterial and fungal cultures are restricted and can only be ordered by educational professionals.
Caution: Although this organism is non pathogenic, Ward’s recommends that you utilize proper aseptic technique when handling all microorganisms.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty