Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ward's® Live Serratia marcescens Cultures: Strain 933 and Strain WCF

Supplier: VWR

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

WARD470179-150 WARD470179-552 WARD470179-148 WARD470179-554
470179-552KT 19.99 USD InStock
470179-552 470179-148 470179-554 470179-150
Ward's® Live Serratia marcescens Cultures: Strain 933 and Strain WCF
Specimens Live Specimens Bacteria - Living Material
Both of these strains of Serratia marcescens are colorless mutants when plated alone, but when grown together, they produce a red pigment.

  • Biosafety Level: 1
  • Canada: Containment Level 2 - PHAC Licence required
  • Motility: Positive
  • Morphology: Rods
  • Gram Reaction: Negative
  • Respiration: Aerobic

All microbial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.

These organisms are offered as ready-to-use tube cultures, as well as in freeze-dried cultures for longer storage. Freeze-dried organisms come with everything you need to rehydrate and culture the organism when you're ready.

Ward’s Science is not affiliated with the American Type Culture Collection and does not provide ATCC numbers for our organisms.

Ordering information: Microbial and fungal cultures are restricted and can only be ordered by educational professionals.

These products contain living or perishable materials and will ship via overnight or second day delivery. Please specify a delivery date during checkout

Caution: While this organism is non-pathogenic, Ward’s recommends that you utilize proper aseptic technique when handling all microorganisms.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty