Would you like to log in again?
Ward's® Live Streptococcus equisimilis Culture - PATHOGEN
Supplier: VWR
WARD470179-166
470179-166EA 13.25 USD InStock
470179-166
Ward's® Live Streptococcus equisimilis Culture - PATHOGEN
Specimens Live Specimens Bacteria - Living Material
Lancefield group C, beta hemolysis. Causes strangles in horses. May be pathogenic in humans. Camp test negative; resistant to bacitracin, does not ferment lactose. 
  • Biosafety Level: 2
  • Motility: Negative
  • Morphology: Cocci in pairs and chains
  • Gram Reaction: Positive
  • Respiration: Aerobic
  • Hemolysis: Beta
All microbial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.
This organism is offered as a ready-to-use tube culture.
Ward’s Science is not affiliated with the American Type Culture Collection and does not provide ATCC numbers for our organisms.
Canada: Containment Level 2 - PHAC Licence required
Ordering information: Microbial and fungal cultures are restricted and can only be ordered by educational professionals.
These products contain living or perishable materials and will ship via overnight or second day delivery. Please specify a delivery date during checkout
Caution: This organism is pathogenic, Ward’s recommends that you utilize proper aseptic technique when handling all microorganisms.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty