Would you like to log in again?
Ward's® Live Streptococcus viridians Culture - PATHOGEN
Supplier: VWR
WARD470179-550 WARD470179-178
470179-178EA 13.25 USD InStock
470179-178 470179-550
Ward's® Live Streptococcus viridians Culture - PATHOGEN
Specimens Live Specimens Bacteria - Living Material
Isolated from human saliva, sputum, and intestine. No Lancefield group shown, alpha hemolysis.
  • Biosafety Level: 2
  • Canada: Containment Level 2 - PHAC Licence required
  • Motility: Negative
  • Morphology: Cocci in chains (long chains in broth)
  • Gram Reaction: Positive
  • Respiration: Aerobic
  • Hemolysis: Alpha
All microbial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.
This organism is offered as a ready-to-use tube culture, as well as in a freeze-dried culture for longer storage. Freeze-dried organisms come with everything you need to rehydrate and culture the organism when you're ready.
Ward’s Science is not affiliated with the American Type Culture Collection and does not provide ATCC numbers for our organisms.
Ordering information: Microbial and fungal cultures are restricted and can only be ordered by educational professionals.
These products contain living or perishable materials and will ship via overnight or second day delivery. Please specify a delivery date during checkout
Caution: This organism is pathogenic, Ward’s recommends that you utilize proper aseptic technique when handling all microorganisms.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty