Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ward's® Moraxella catarrhalis

Supplier: VWR
WARD470179-128 WARD470179-494
470179-494EA 19.99 USD InStock
470179-494 470179-128
Ward's® Moraxella catarrhalis
Specimens Live Specimens Bacteria - Living Material
Normal flora of respiratory tract. Beta-lactamase positive. Also known by the name Branhamella catarrhalis

  • Biosafety Level: 1
  • Canada: Containment Level 2 - PHAC Licence required
  • Motility: Negative
  • Morphology: Cocci in pairs
  • Gram Stain: Negative with a tendency to resist coloration
  • Respiration: Aerobic

Normal flora of respiratory tract. Beta-lactamase positive. Nonmotile. Cocci in pairs. Gram negative reaction; tendency to resist decoloration. Aerobic. Incubation temperature 37°C. Blood agar growth medium.

Packaged in a 16 x 125 mm tubes. Call our Customer Service for availability of demo plates and other special orders.

All microbial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.

This organism is offered as a ready-to-use tube culture, as well as in a freeze-dried culture for longer storage. Freeze-dried organisms come with everything you need to rehydrate and culture the organism when you're ready.

Ward’s Science is not affiliated with the American Type Culture Collection and does not provide ATCC numbers for our organisms.

Ordering information: Microbial and fungal cultures are restricted and can only be ordered by educational professionals.

These products contain living or perishable materials and will ship via overnight or second day delivery. Please specify a delivery date during checkout

Caution: Though this organism is not pathogenic, Ward’s recommends that you utilize proper aseptic technique when handling all microorganisms.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty