Would you like to log in again?
Ward's® Live Lactobacillus casei Culture
Supplier: VWR
WARD470179-124 WARD470179-524
470179-124EA 18.95 USD InStock
470179-124 470179-524
Ward's® Live Lactobacillus casei Culture
Specimens Live Specimens Bacteria - Living Material
Isolated from milk and cheese products. Can be used to ferment milk. 
  • Biosafety Level: 1
  • Motility: Negative
  • Morphology: Rods in chains
  • Gram Stain: Positive
  • Respiration: Facultatively anaerobic, sometimes microaerophilic
All microbial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.
This organism is offered as a ready-to-use tube culture, as well as in a freeze-dried culture for longer storage. Freeze-dried organisms come with everything you need to rehydrate and culture the organism when you're ready.
Ward’s Science is not affiliated with the American Type Culture Collection and does not provide ATCC numbers for our organisms.
Ordering information: Microbial and fungal cultures are restricted and can only be ordered by educational professionals.
These products contain living or perishable materials and will ship via overnight or second day delivery. Please specify a delivery date during checkout
Caution: This product is for educational use only, do not use this organism to ferment milk that will be consumed.
Though this organism is not pathogenic, Ward’s recommends that you utilize proper aseptic technique when handling all microorganisms.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty