Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ward's® Live Staphylococcus aureus Coagulase-Negative Culture (Pathogenic)

Supplier: VWR
Ward's®
WARD470179-160 WARD470179-562
470179-562EA 19.99 USD InStock
470179-562 470179-160
Ward's® Live Staphylococcus aureus Coagulase-Negative Culture (Pathogenic)
Specimens Live Specimens Bacteria - Living Material
This yellow, shiny organism is known to cause wound infections and more famously, toxic shock syndrome.

  • Biosafety Level: 2
  • Canada: Containment Level 2 - PHAC License required
  • Gram Stain: Positive
  • Morphology: Cocci in grape like clusters
  • Coagulase: Negative
  • Catalase: Positive
  • Motility: Negative
  • Respiration: Aerobic
  • Chromogenesis: Yes (yellow-to-orange)

All bacterial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.

This organism is offered as a ready-to-use agar slant tube culture, as well as in a freeze-dried culture for longer storage.

Freeze-dried organisms come with everything you need to rehydrate and culture the organism when you're ready.

Ordering information: Bacterial and fungal cultures are restricted and can only be ordered by educational and research professionals.

Ward's Science is not affiliated with the American Type Culture Collection and does not provide corresponding ATCC numbers for our organisms.

Delivery information: These products contain living or perishable materials and will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.

Caution: This organism is pathogenic. Ward's adheres to the Biosafety Guidelines for Microbiological and Biomedical Laboratories, as published by the National Institutes of Health.

As with all organisms, Ward's recommends that you utilize proper aseptic technique when handling all microorganisms.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty