Would you like to log in again?
Ward's® Bacteria Forms Set
Supplier: VWR
470179-610KT 42.6 USD InStock
470179-610
Ward's® Bacteria Forms Set
Specimens Live Specimens Bacteria - Living Material
Includes Examples of the Three Basic Morphological Types
View all three types of bacteria individually, then compare their different structures, with the selection of specimens in this set. Tube cultures: Bacillus subtilis (rod), Rhodospirillum rubrum (spirillum), and Kocuria rhizophila (coccus). The set of three is available with or without an Explano-Mount, which is a microscope slide with an illustrated guide that highlights all the important information on the slide and the Working with Bacteria manual. Set of three.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty