Would you like to log in again?
Mucor hiemalis (+)
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
470179-712EA 18.95 USD InStock
470179-712
Mucor hiemalis (+)
Specimens Live Specimens Fungi - Living Material
Zygomycete. Dung Mold. Mating strain; cross with Mucor heimalis (-) for zygospore formation. Incubation temperature 25°C. Potato dextrose agar growth medium
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty