Would you like to log in again?
Ward's® Live Chlamydomonas moewusii Cultures
Supplier: VWR
WARD470179-732 WARD470179-738 WARD470179-734 WARD470179-736 WARD470179-828
470179-732EA 9.5 USD InStock
470179-732 470179-736 470179-734 470179-828 470179-738
Ward's® Live Chlamydomonas moewusii Cultures
Specimens Live Specimens Protists - Living Material
Demonstrates isogamous reproduction where gametes are similar in appearance and behavior. Make lectures and labs on protist reproduction clearer with our specially selected cultures that illustrate various reproductive phenomona.
  • Both mating types (+) and (-) available
  • Bacteria-free (Axenic) cultures available for both mating types
This green alga is a unicellular flagellate commonly found in damp soil, and aquatic environments . Each cell has 2 flagella and a large eyespot that allow it to sense light.
Chlamydomonas moewusii can only survive in high levels of light, while C. reinhartdii (also available) can also survive in the dark if it is provided organic carbon.
Genus: Chlamydomonas Species: moewusii
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty