Would you like to log in again?
Coccolithophora
Supplier: VWR
COCCO30/35
470179-912EA 9.5 USD InStock
470179-912
Coccolithophora
Specimens Live Specimens Protists - Living Material
Marine. Unicellular; contains same pigments as other Golden Algae but contains haptonema, unique to Haptophyceae. Coccoid, from coccoliths. Marine medium. Characterized by golden yellow plastids.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty