Would you like to log in again?
Wandering Jew
Supplier: CHASE GREENHOUSES, INC.
867300
470180-026EA 7.35 USD InStock
470180-026
Wandering Jew
Specimens Live Specimens Plants - Living Material
Tradescantia virginiana or similar. Monocot. This hardy, easy-to-grow ornamental plant can be used for studying meiotic division in mother cells of pollen. Cytoplasmic flows can be seen in cell hairs on staminate filaments. It is native to tropical and subtropical areas of the Americas and is supplied in a pot with care instructions. Size: 6".
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty