Would you like to log in again?
Venus’ Flytrap
Supplier: ROBERT CRESPO
VFT-25-POT-3INCH
470180-030EA 19.3 USD InStock
470180-030
Venus’ Flytrap
Specimens Live Specimens Plants - Living Material
Dionaea sp. Dicot. A fascinating plant to observe, the leaves close around anything that brushes against the inner hairs. Trapped insects are digested by enzymes the leaves secrete. It is native to North America, especially the Carolinas and prefers acidic conditions such as a bog terrarium. It comes supplied in a pot with care instructions. Size: 2– 4".
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty