Would you like to log in again?
Ward's® Live Egeria sp. Aquatic Plants
Supplier: Avantor
Ward's Ward's®
WARD470354-358 WARD470180-036 WARD470198-312 WARD470175-814 WARD470136-656 WARD470136-600 WARD470136-612 WARD470136-610 WARD470183-752 WARD470225-402 WARD470176-598 WARD470225-398 WARD470136-594 WARD470136-598 WARD470180-034 WARD470180-038 WARD470183-806
470183-806EA 18.95 USD InStock
470183-806 470136-656 470136-612 470225-398 470136-600 470225-402 470180-034 470136-610 470180-036 470175-814 470136-598 470180-038 470136-594 470354-358 470176-598 470198-312 470183-752
Ward's® Live Egeria sp. Aquatic Plants
Specimens Live Specimens Plants - Living Material
An excellent aquarium oxygenator, great for teaching respiration, photosynthesis, and plasmolysis.
  • Can be rooted in substrate, or float on the surface
  • Sprigs or tips are useful to demonstrate photosynthesis or plasmolysis
  • Tips have fresh growth to demonstrate streaming protoplasm and osmosis
Egeria najas and Egeria densa (formally Elodea densa) are popular aquarium plants that help aerate the tank to help supply oxygen to fish and pull unwanted nitrogen from the water. They have the added benefit of being a very useful plant to teach numerous lessons in the classroom.
As Egeria densa is considered a highly invasive species, it is highly recommended to choose Egeria najas instead, if it is available in your state or territory. This closely related plant looks very similar and can be used for all the same purposes as Egeria densa. Also offered are other alternative aquatic plants that are native to the United States such as Elodea canadensis, and Ceratophyllum demersum. Do not hesitate to reach out to our technical support team if you need any assistance in picking what plant would work best for your own unique needs.
Ordering information: These specimens are considered invasive and are restricted in certain states and territories, and therefore your order may be held for review by a regulatory department prior to shipping. Please give additional lead time when requesting delivery to allow for this review. If you need assistance picking an option available in your state or territory, please reach out to our Science Help technical support team for assistance.
Delivery information: These specimens are perishable and will ship via two day delivery. Please provide a requested delivery date.
Packaging: Plants are provided in waterproof plastic bags that keep plants both fresh and healthy. Care instructions available online.
Caution: Do not release invasive specimens into the wild.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty