Would you like to log in again?
Ward's® Live Elodea canadensis Aquatic Plants
Supplier: VWR
Ward's
WARD470180-094 WARD470180-090 WARD470177-082 WARD470180-088 WARD470180-092
470180-090PK 29.95 USD InStock
470180-090 470180-092 470180-094 470177-082 470180-088
Ward's® Live Elodea canadensis Aquatic Plants
Specimens Live Specimens Plants - Living Material
An excellent aquarium oxygenator, Elodea canadensis is great for teaching photosynthesis and plasmolysis.
  • Sprigs or tips are useful to demonstrate photosynthesis or plasmolysis
  • Float in your aquarium as an oxygenating hiding place for small animals
Excellent alternative if Elodea densa or Egeria najas are restricted in your state. This aquatic monocot is slightly smaller than Elodea densa and does not grow as straight sprigs, but curls into bundles.
Tips are not recommended for aquarium use. Instead, sprigs may be floated in aquaria as a food source or hiding place for fish and snails.
Ordering information: May be restricted in some states, check with your state's Department of Agriculture for restrictions. Elodea densa or Egeria najas are also available as alternatives.
Delivery information: These specimens are perishable and will ship via 2-day delivery. Please provide a requested delivery date.
Packaging: Plants are provided in waterproof, plastic bags that keep plants both fresh and healthy. Care instructions available online.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty