Would you like to log in again?
Ward's® Live Brown Planaria (Dugesia dorotocephala)
Supplier: VWR
WARD470176-558 WARD470176-560 WARD470180-236
470176-560EA 39.95 USD InStock
470176-560 470180-236 470176-558
Ward's® Live Brown Planaria (Dugesia dorotocephala)
Specimens Live Specimens Animals - Living Material
This is the most widely studied Turbellarian, a free-living flatworm commonly used in science fair projects.
  • Students Research Structure, Feeding Habits, Responses to Environmental Stimuli, and Regeneration
  • Easy-To-Maintain
  • Planaria Feed on Animal Matter
  • Water Must Be Changed After Feeding
Brown planaria are easy to maintain. They feed on animal matter such as liver and are up to 1/4" in size.
Ordering information: These products contain living or perishable materials. Please specify a delivery date.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty