Would you like to log in again?
Soil Nematodes
Supplier: CAROLINA BIOL SUPPLY CO SE
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
WARD470180-246 WARD470180-248
470180-246EA 19.95 USD InStock
470180-246 470180-248
Soil Nematodes
Specimens Live Specimens Animals - Living Material
Examine the morphological characteristics of typical soil nematodes with the specimens in our well-populated culture.
  • Keeps for up to two months if refrigerated
  • Examines morphological characteristics
  • Well-populated culture
Examine the morphological characteristics of typical soil nematodes with the specimens in our well-populated culture. Refrigerate the culture at 48°F and it should keep for up to two months. Culture instructions are included. Size: 0.5–5 mm.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty