Would you like to log in again?
Rhabditis
Supplier: CAROLINA BIOL SUPPLY CO SE
13-3258
470180-250EA 13.45 USD InStock
470180-250
Rhabditis
Specimens Live Specimens Animals - Living Material
Rhabditis sp. Easily grown on potato medium and easy to maintain, Rhabditis can be used as food for predatory fungi such as Arthrobotrys. You will receive enough materials for 35 students and culture instructions.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty