Would you like to log in again?
Aeolosoma
Supplier: VWR
L29
470180-288EA 27.95 USD InStock
470180-288 470180-286
Aeolosoma
Specimens Live Specimens Aquaria - Living Material
Aeolosoma sp. A small, freshwater Oligochaete, Aeolosoma is characterized by colorful oil droplets in its body. It’s easy to culture large quantities of this microscopic organism for use in regeneration experiments: Simply add hay and wheat grains to hay medium. Culture instructions are included. Size: 2–3 mm.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty