Would you like to log in again?
Ward's® Live Termite (Zootermopsis sp.)
Supplier: VWR
WARD470180-354
470180-354PK 39.95 USD InStock
470180-354
Ward's® Live Termite (Zootermopsis sp.)
Specimens Live Specimens Animals - Living Material
This species of termite is very large, and you can easily observe the distinct differences of the several roles that exist in their eusocial colonies.
  • Exhibit "Caste" system
  • Excellent model for symbiosis
  • Extremely easy to maintain in the laboratory or classroom
  • No reproductive termites are sent (we do not send queens)
One of the two different genera of termites available at Ward's, Zootermopsis are known as dampwood termites. They are commonly located in the Pacific northwest and southern British Columbia where they make their homes in rotting logs and other dark, moist wooden areas. They have a symbiotic relationship with a flagellate protozoa in their gut that helps them digest the cellulose in the wood.
Ordering information: These products contain living or perishable materials. No reproductive termites are sent with the culture. We do not send queens.
Delivery information: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Ward's holds valid USDA permits allowing us to ship to most continental US states. Please call Customer Service for permit details.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty