Would you like to log in again?
Ward's® Live Milkweed Bugs and Eggs (Oncopeltus fasciatus. )
Supplier: VWR
WARD470176-486 WARD470180-356 WACP470189-340
470180-356PK 18.95 USD InStock
470180-356 470189-340 470176-486
Ward's® Live Milkweed Bugs and Eggs (Oncopeltus fasciatus. )
Specimens Live Specimens Animals - Living Material
Named for their food preference of milkweed seeds, these colorful Hemipterans demonstrate simple metamorphosis and sucking mouthparts.
  • Complete metamorphosis
  • Sucking mouthparts
Ordering information: These products contain living or perishable materials
Delivery information: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty