Would you like to log in again?
Ward's® Live Silkworm Eggs (Bombyx mori)
Supplier: VWR
WARD470180-380 WARD470180-382
470180-380PK 24.95 USD InStock
470180-380 470180-382
Ward's® Live Silkworm Eggs (Bombyx mori)
Specimens Live Specimens Animals - Living Material
When hatched, the small, gray larvae are the traditional source for high-quality silk.
  • They can remain refrigerated for up to a year, allowing for easy storage and access
  • Eggs will hatch in 10-14 days if kept at 75-85°F
Over thousands of years of domestication and selective breeding the modern silkworm has lost its ability to fly. Eggs, each roughly the size of a pinhead, come glued onto a petri dish.
Ordering information: These products contain living or perishable materials.
Delivery information: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty