Would you like to log in again?
Ward's® Live Sarcophaga Bullata Fly Larvae and Pupae
Supplier: VWR
WARD470180-392 WARD470180-472 WARD470180-394 WARD470177-084
470180-392EA 21.95 USD InStock
470180-392 470180-394 470177-084 470180-472
Ward's® Live Sarcophaga Bullata Fly Larvae and Pupae
Specimens Live Specimens Animals - Living Material
Sarcophaga bullata are gray flesh flies. These flies are reared on decaying flesh and pupate quickly, within 5 to 7 days from birth, to illustrate complete metamorphosis in Diptera.
The (uninfected) pupae can serve as the host organism for emerging Nasonia larvae. The pupae can be stored under refrigeration for several weeks, effectively halting their life cycle and assuring a fresh supply of food for the Nasonia larvae.
Ordering information: Please specify a delivery date during checkout.
To request a specific instar, please contact us ten days in advance.
Delivery information: These products contain living or perishable materials and will ship via overnight or 2nd day delivery.
Packaging: A large, vigorous culture is shipped in a shatterproof plastic vial.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty