Would you like to log in again?
Ward's® Live Drosophila melanogaster - Chromosome I Mutants
Supplier: VWR
WARD470180-412 WARD470180-450 WARD470194-748 WARD470176-756 WARD470176-782 WARD470194-742 WARD470194-736 WARD470194-740 WARD470180-416 WARD470180-418
470194-736EA 9.95 USD InStock
470194-736 470194-748 470180-412 470176-782 470194-742 470180-416 470194-740 470180-418 470180-450 470176-756
Ward's® Live Drosophila melanogaster - Chromosome I Mutants
Specimens Live Specimens Animals - Living Material
Drosophila, the common fruit fly is widely used in genetic studies. Ward's offers a large number of mutations to help you demonstrate complex genetics concepts.
  • Help your students to understand sex-linked, dominant and recessive modes of inheritance
  • Easy to maintain, with short generation times
  • Our cultures come with our satisfaction guarantee for genetic purity and excellent quality
Help your students to understand sex-linked, dominant and recessive modes of inheritance, using any of the many mutant fly lines we have available. All cultures come with a mixture of male and female flies of various ages to ensure culture health.
White (w) is a recessive mutation that causes the flies to have eyes that are un-pigmented, appearing white in color, rather than the bright red of a wild-type fly.
Apricot (a) is a recessive mutation that causes the flies to have eyes that are apricot in color, rather than the bright red of a wild-type fly.
Ruby (rb) is a recessive mutation that causes the flies to have eyes that are dark red, ruby-colored.
Vermillion (v) is a recessive mutation that causes the flies to have eyes that are bright red color.
Bar (B) is a dominant mutation that causes the flies to have reduced numbers of eye facets, so their eyes are thinner than normal. A wild-type fly's eye is very round, these are 'bar-like'.
Miniature (m) is a recessive mutation that causes flies to have wings that are smaller than normal. They are only slightly longer than the abdomen, whereas a wild-type fly's wings stick out much farther. Miniature' flies wings have normal proportions, but appear dark gray and less transparent than a wild-type fly.
Forked (f) is a recessive mutation that causes the flies to have body-bristles that are forked at the ends, rather than straight and pointed as in a wild-type fly.
White-Yellow (w; y) is combination of two recessive mutations causing the flies to have white eyes and yellow bodies.
White-Miniature-Yellow (w; m; y) is a combination of three recessive mutations, causing the flies to have white eyes, smaller than usual wings, and yellow bodies.
Yellow-Forked-Attached-White Males have white eyes, a wild-type body color and wings. Females have wild-type eyes, a yellow body with forked bristles, and normal wings.
Delivery information: These products contain living or perishable materials and must ship via overnight or 2-day transit. Please specify a delivery date at checkout.
Packaging: Cultures are shipped with Drosophila medium in a shatterproof vial. The pupae will usually appear within ten days of the ship date.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty