Would you like to log in again?
Drosophila mojavensis
Supplier: VWR
CALMOJA
470180-452EA 10.25 USD InStock
470180-452
Drosophila mojavensis
Specimens Live Specimens Animals - Living Material
Long life cycle (approximately one month). Useful for comparative population studies and for preparing salivary gland chromosome squashes.
Recommended by the Carnegie Institute
Mature Drosophila are cultured in our own labs, and are marked with the culture initiation date. Culture is shipped in our instant medium in a shatterproof vial. The pupae will usually appear within ten days of the ship date. You will also receive WARD’S Drosophila Culture Guide. Note: We can supply any of our single strains in the larger quantity for 100 students. Call our Customer Service department for details.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty