Would you like to log in again?
Ward's® Live Jellyfish (Cassiopeia sp.)
Supplier: VWR
WARD470180-490
470180-490KT 47.95 USD InStock
470180-490
Ward's® Live Jellyfish (Cassiopeia sp.)
Specimens Live Specimens Aquaria - Living Material
Pack of 3 Jellyfish (Cassiopeia sp.) Students can observe symbiotic organisms called zooxanthellae (minute flagellates) swimming with the jellyfish in an inverted position, with tentacles overhead.`
  • Symbiotic relationship with Zooxanthellae
  • Saltwater specimen
Ordering information: These products contain living or perishable materials
Delivery information: Specimens will ship via overnight delivery. Additional charges will apply.
Please specify a delivery date during checkout.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty