Would you like to log in again?
Ward's® Live Sea Peach (Halocynthia pyriformis)
Supplier: VWR
WARD470180-498
470180-498EA 29.95 USD InStock
470180-498
Ward's® Live Sea Peach (Halocynthia pyriformis)
Specimens Live Specimens Aquaria - Living Material
This brightly colored filter feeder, also known as a "sea squirt" is suitable for marine aquariums.
  • Exhibits interesting feeding behavior
  • Makes an excellent addition to any saltwater tank
This Tunicate is a primitive Chordate that displays a notochord in the larval stage that disappears as it becomes an adult.
Ordering information: These products contain living or perishable materials
Delivery information: Specimens will ship via overnight delivery. Additional charges will apply.
Please specify a delivery date during checkout.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty