Would you like to log in again?
Ward's® Live Grassfrogs (Rana pipiens)
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
WARD470180-586 WARD470180-602 WARD470180-588 WARD470180-590 WARD470180-604 WARD470180-598
470180-598PK 60.95 USD InStock
470180-598 470180-586 470180-588 470180-602 470180-604 470180-590
Ward's® Live Grassfrogs (Rana pipiens)
Specimens Live Specimens Animals - Living Material
Grass frogs are low maintenance and seasonally available, typically from September to July.
  • Great for In-Class Observation
  • Low-Maintenance
  • Also Known as Leopard Frogs
Keep in an aquarium or terrarium with both wet and dry areas and feed crickets and/or earthworms.
Ordering information: This kit includes living or perishable materials. They may not be available in all states. Check with your local department of agriculture.
Delivery information: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
CANADIAN CUSTOMERS: Please note that due to Canadian import restrictions, frogs can no longer be packed with sphagnum moss.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty