Would you like to log in again?
Ward's® Live Bullfrogs (Rana catesbeiana)
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
WARD470180-616 WARD470180-614 WARD470176-296 WARD470189-248
470176-296PK 304.95 USD InStock
470176-296 470180-614 470180-616 470189-248
Ward's® Live Bullfrogs (Rana catesbeiana)
Specimens Live Specimens Animals - Living Material
This large predatory frog will eat any other organism smaller than itself, including smaller bullfrogs; keep similar-sized frogs together to avoid cannibalism.
  • Large specimens
  • Excellent for observation in large aquaria.
This large predatory frog will eat any other organism smaller than itself, including smaller bullfrogs; keep similar-sized frogs together to avoid cannibalism. Frogs are 5" or larger in size.
Ordering Information: Contact your Sales Representative or call Customer Service to check availability.
Ordering information: These products contain living or perishable materials
This product is available to US customers only.
Delivery information: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty