Would you like to log in again?
Defibrinated, Prepared Animal Blood
Supplier: LAMPIRE BIOLOGICAL LABS, INC.
7233401 7239001 7230801
470180-678EA 124.95 USD InStock
470180-678 470177-372 470180-682
Defibrinated, Prepared Animal Blood
Blood
Products are for Laboratory Use Only.
  • Aseptically collected from healthy donor animals
  • Mechanically defibrinated - no coagulants
  • Quality tested, and lot controlled
  • Keep refridgerated
  • 3 week shelf life
These various blood samples can be used to study the osmotic properties of blood cells, as well as for other basic hematology studies, or to make your own microbiological blood agar.
Ordering information: Our blood samples are only sold to accredited colleges and laboratories. These 100mL blood samples are collected aseptically from healthy donor animals. The blood is defibrinated mechanically without the addition of anticoagulants. Each lot is inspected for various physical characteristics and tested for hematocrit levels and presence of bioburden prior to release. Test summary is available upon request, please make note of your lot-code before calling customer service. Shelf life: Stable for up to three (3) weeks from bleed date under recommended storage (2-8°C). The actual period of suitability for use may be longer or shorter according to the specific application.
Delivery information: Specimens will ship via overnight for delivery. Please specify a delivery date during checkout. Please allow 2-3 weeks for delivery for Horse Blood, or for large orders.
Caution: These products are for laboratory use only.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty