Would you like to log in again?
Prepared Blood Agar Media
Supplier: CULTURE MEDIA &SUPPLIES INC SE
Avg. Ratings: 5.0 (1 Review)
REVIEWS (1)
Total Ratings: 1
Avg. Ratings: out of 5
Sort all by:
WARD470180-642 880901
470180-642CS 271.92 USD InStock
470180-642 470180-680
Prepared Blood Agar Media
Media for Microbiology Dehydrated Media for Microbiology
Tryptic Soy Agar base with 5% Sheep Blood
  • Excellent for cultivation of fastidious organisms
  • Plate your organism onto Blood agar plates to test for hemolytic reaction
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (1)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty